Invitrogen
Manufacturer Code:PA572189
Catalog # PIPA572189
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 547-580 amino acids from the C-terminal region of human SV2C |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Synaptic vesicle glycoprotein 2C; Synaptic vesicle glycoprotein 2C SV2C KIAA1054; Synaptic vesicle protein 2C
Gene Aliases: Sv2c
UniProt ID: (Rat) Q9Z2I6
Entrez Gene ID: (Rat) 29643
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.