Novus Biologicals
Manufacturer Code:NBP30505220UL
Catalog # NB096271
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 460-580 of human SV2C (NP_055794.3). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KIAA1054 MGC118874 MGC118875 MGC118876 MGC118877 synaptic vesicle glycoprotein 2C synaptic vesicle protein 2C; Synaptic vesicle glycoprotein 2C; synaptic vesicle protein 2C
Gene Aliases: KIAA1054; SV2C
UniProt ID: (Human) Q496J9
Entrez Gene ID: (Human) 22987
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.