Invitrogen
Manufacturer Code:PA518581
Catalog # PIPA518581
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (TVELYPSDKRTTA) corresponding to the internal amino acids of SV2A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Calcium regulator SV2A; SV2A synaptic vesicle glycoprotein 2A KIAA0736 SV2 synaptic vesicle glycoprotein 2; Synaptic vesicle glycoprotein 2A; Synaptic vesicle protein 2; synaptic vesicle protein 2A; synaptic vesicle transmembrane transporter (SV2) RNA
Gene Aliases: AI746429; KIAA0736; mKIAA0736; PSEC0174; SV2; SV2A
UniProt ID: (Human) Q7L0J3, (Mouse) Q9JIS5, (Rat) Q02563
Entrez Gene ID: (Human) 9900, (Mouse) 64051, (Rat) 117559
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.