Invitrogen
Manufacturer Code:PA5113833
Catalog # PIPA5113833
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human SV2A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AI746429 Calcium regulator SV2A Kiaa0736 mKIAA0736 PSEC0174 Sv2 SV2A synaptic vesicle glycoprotein 2 a Synaptic vesicle glycoprotein 2A Synaptic vesicle protein 2 synaptic vesicle protein 2A synaptic vesicle transmembrane transporter (SV2) RNA; Calcium regulator SV2A; Synaptic vesicle glycoprotein 2A; Synaptic vesicle protein 2; synaptic vesicle protein 2A
Gene Aliases: AI746429; KIAA0736; mKIAA0736; PSEC0174; SV2; SV2A
UniProt ID: (Human) Q7L0J3, (Mouse) Q9JIS5
Entrez Gene ID: (Human) 9900, (Mouse) 64051
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.