Invitrogen
Manufacturer Code:PA584109
Catalog # PIPA584109
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human KMT5C |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: [histone H4]-lysine20 N-methyltransferase KMT5B; [histone H4]-N-methyl-L-lysine20 N-methyltransferase KMT5B; Histone-lysine N-methyltransferase KMT5C; histone-lysine N-methyltransferase SUV420H2; lysine (K)-specific methyltransferase 5C; Lysine N-methyltransferase 5C; Lysine-specific methyltransferase 5C; MGC2705 SUV420H2; Su(var)4-20 homolog 2; Suppressor of variegation 4-20 homolog 2; suv4-20h2
Gene Aliases: KMT5C; PP7130; SUV420H2
UniProt ID: (Human) Q86Y97
Entrez Gene ID: (Human) 84787
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.