Invitrogen
Manufacturer Code:PA5115484
Catalog # PIPA5115484
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human KMT5C/SUV4-20h2 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: [histone H4]-lysine20 N-methyltransferase KMT5B; [histone H4]-N-methyl-L-lysine20 N-methyltransferase KMT5B; BC024816 Histone-lysine N-methyltransferase KMT5C histone-lysine N-methyltransferase SUV420H2 KMT5C lysine (K)-specific methyltransferase 5C lysine methyltransferase 5C lysine N-methyltransferase 5C Lysine-specific methyltransferase 5C PP7130 Su(var)4-20 homolog 2 suppressor of variegation 4-20 homolog 2 suppressor of variegation 4-20 homolog 2 (Drosophila) SUV420H2 suv4-20h2; Histone-lysine N-methyltransferase KMT5C; histone-lysine N-methyltransferase SUV420H2; lysine (K)-specific methyltransferase 5C; Lysine N-methyltransferase 5C; Lysine-specific methyltransferase 5C; Su(var)4-20 homolog 2; Suppressor of variegation 4-20 homolog 2; suv4-20h2
Gene Aliases: BC024816; KMT5C; PP7130; Suv4-20h2; SUV420H2
UniProt ID: (Human) Q86Y97, (Mouse) Q6Q783, (Rat) P0C2N6
Entrez Gene ID: (Human) 84787, (Mouse) 232811, (Rat) 308345
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.