Invitrogen
Manufacturer Code:PA569767
Catalog # PIPA569767
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ChIP assay (ChIP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human SUV420H1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: [histone H4]-lysine20 N-methyltransferase KMT5B; [histone H4]-N-methyl-L-lysine20 N-methyltransferase KMT5B; C630029K18Rik; Histone-lysine N-methyltransferase KMT5B; histone-lysine N-methyltransferase SUV420H1; lysine (K)-specific methyltransferase 5B; Lysine N-methyltransferase 5B; Lysine-specific methyltransferase 5B; Su(var)4-20 homolog 1; Suppressor of variegation 4-20 homolog 1; Suppressor of variegation 4-20 homolog 1 (Drosophila) CGI-85 CGI85 KMT5B MGC118906 MGC118909 MGC21161 MGC703; suppressor of variegation 4-20 protein-like 1; suv4-20h1
Gene Aliases: AA117471; C630029K18Rik; CGI-85; CGI85; KMT5B; si:dkey-12e7.2; Suv4-20h1; SUV420H1; wu:fb97g06; wu:fi57g03; zgc:103527
UniProt ID: (Human) Q4FZB7, (Bovine) Q29RP8, (Rat) P0C2N5, (Mouse) Q3U8K7, (Zebrafish) Q5U3H2
Entrez Gene ID: (Human) 51111, (Horse) 100053438, (Bovine) 767828, (Rabbit) 100340037, (Dog) 483690, (Guinea pig) 100727663, (Rat) 361688, (Mouse) 225888, (Zebrafish) 572849
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.