Novus Biologicals
Manufacturer Code:NBP171812
Catalog # NBP171812
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen for this product maps to a region between residue 686 and 736 of human Suppressor of Var1 3-like 1 using the numbering given in entry NP_003162.2 (GeneID 6832). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP-dependent RNA helicase SUPV3L1 mitochondrial EC 3.6.1 EC 3.6.4.13 suppressor of var1 (S.cerevisiae) 3-like 1 Suppressor of var1 3-like protein 1 suppressor of var1 3-like 1 (S. cerevisiae) SUV3-like protein 1 SUV3suppressor of var1 3-like 1(SUV3); ATP-dependent RNA helicase SUPV3L1, mitochondrial; Suppressor of var1 3-like protein 1; suppressor of var1, 3-like 1; suppressor of var1, 3-like 1(SUV3); SUV3-like helicase; SUV3-like protein 1
Gene Aliases: SUPV3L1; SUV3
UniProt ID: (Human) Q8IYB8
Entrez Gene ID: (Human) 6832
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.