Invitrogen
Manufacturer Code:PA577031
Catalog # PIPA577031
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human SUV39H2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ23414 H3 K9 HMTase 2 H3-K9-HMTase 2 Histone H3 K9 methyltransferase 2 Histone H3-K9 methyltransferase 2 Histone lysine N methyltransferase H3 lysine 9 specific 2 Histone lysine N methyltransferase SUV39H2 Histone-lysine N-methyltransferase SUV39H2 KMT1B Lysine N methyltransferase 1B Lysine N-methyltransferase 1B sSuppressor of variegation 3 9 homolog 2 (Drosophila) Su(var)3 9 homolog 2 Su(var)3-9 homolog 2 Suppressor of variegation 3 9 homolog 2 Su; H3-K9-HMTase 2; Histone H3-K9 methyltransferase 2; Histone-lysine N-methyltransferase SUV39H2; Lysine N-methyltransferase 1B; Su(var)3-9 homolog 2; Suppressor of variegation 3-9 homolog 2; Suv39h histone methyltransferase
Gene Aliases: 4930507K23Rik; AA536750; D030054H19Rik; D2Ertd544e; KMT1B; SUV39H2
UniProt ID: (Human) Q9H5I1, (Mouse) Q9EQQ0
Entrez Gene ID: (Human) 79723, (Mouse) 64707
Molecular Function:
DNA binding protein
methyltransferase
nucleic acid binding
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.