Invitrogen
Manufacturer Code:PA518115
Catalog # PIPA518115
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ChIP assay (ChIP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (CKCGAVTCRGYLN) corresponding to the C-terminus amino acids of SUV39H2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: histone-lysine N-methyltransferase SUV39H2; SUV39H2 suppressor of variation 3-9 (Drosophila) homolog 2 (Drosophila) FLJ23414 FLJ23414 RP11-2K17.2 KMT1 uppressor of variegation 3-9 homolog 2 (Drosophila) KMT1B suppressor of variegation 3-9 homolog 2
Entrez Gene ID: (Rat) 364785
Molecular Function:
DNA binding protein
methyltransferase
nucleic acid binding
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.