R&D Systems
Manufacturer Code:FAB90561R025
Catalog # FB90561R025
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 944827 |
Immunogen | Human embryonic kidney cell line HEK293-derived human SUSD2 Met1-Ala785 Accession No. Q9UGT4 |
Conjugate | Alexa Fluor 647 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8°C as supplied. |
Mouse Monoclonal Antibody
Protein Aliases: BK65A6.2 FLJ22778 Sushi domain (SCR repeat) containing sushi domain containing 2 sushi domain-containing protein 2; Sushi domain (SCR repeat) containing; Sushi domain-containing protein 2; testicular tissue protein Li 190
Gene Aliases: BK65A6.2; SUSD2
UniProt ID: (Human) Q9UGT4
Entrez Gene ID: (Human) 56241
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.