R&D Systems
Manufacturer Code:MAB9056SP
Catalog # MAB9056SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 944812 |
Immunogen | Human embryonic kidney cell line HEK293-derived human SUSD2 Met1-Ala785 Accession # Q9UGT4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: BK65A6.2 FLJ22778 Sushi domain (SCR repeat) containing sushi domain containing 2 sushi domain-containing protein 2; Sushi domain (SCR repeat) containing; Sushi domain-containing protein 2; testicular tissue protein Li 190
Gene Aliases: BK65A6.2; SUSD2
UniProt ID: (Human) Q9UGT4
Entrez Gene ID: (Human) 56241
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.