Invitrogen
Manufacturer Code:PA569676
Catalog # PIPA569676
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the N terminal region of human SURF4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ERV29 surface 4 integral membrane protein surfeit locus protein 4; surface 4 integral membrane protein; surfeit gene 4; Surfeit locus protein 4
Gene Aliases: AL033340; AL033373; ERV29; SURF-4; SURF4; zgc:65806; zgc:77007
UniProt ID: (Human) O15260, (Bovine) A7YY49, (Mouse) Q64310, (Zebrafish) Q7SZQ7
Entrez Gene ID: (Pig) 100623620, (Dog) 491269, (Human) 6836, (Bovine) 526045, (Guinea pig) 100715812, (Mouse) 20932, (Rat) 619346, (Zebrafish) 394102
Molecular Function: oxidase oxidoreductase transmembrane receptor regulatory/adaptor protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.