Novus Biologicals
Manufacturer Code:NBP144374
Catalog # NBP144374
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 18 amino acid peptide from N-Terminus of human SURF4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ERV29 FLJ22993 MGC102753 SURF-4 surface 4 integral membrane protein surfeit 4 surfeit locus protein 4; surface 4 integral membrane protein; Surfeit locus protein 4
Gene Aliases: ERV29; SURF-4; SURF4
UniProt ID: (Human) O15260
Entrez Gene ID: (Human) 6836
Molecular Function:
oxidase
oxidoreductase
transmembrane receptor regulatory/adaptor protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.