Proteintech
Manufacturer Code:153791AP150UL
Catalog # 50-173-0189
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SURF1 Fusion Protein Ag3959 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: SURF 1 SURF1 surfeit 1 Surfeit locus protein 1; Surfeit locus protein 1
Gene Aliases: 0610010F23Rik; Ab1-205; CMT4K; SURF-1; SURF1
UniProt ID: (Human) Q15526, (Rat) Q9QXU2, (Mouse) P09925
Entrez Gene ID: (Human) 6834, (Rat) 64463, (Mouse) 20930
Molecular Function: oxidase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.