Novus Biologicals
Manufacturer Code:NBP259320UV
Catalog # NBP259320UV
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S289-16 |
Immunogen | Fusion protein amino acids 1548-1582 (cytoplasmic C-terminus) of rat SUR1 |
Conjugate | DyLight 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: ABC36 ATP-binding cassette sub-family C (CFTR/MRP) member 8 HHF1Sulfonylurea receptor 1 HI PHHIHRINS sulfonylurea receptor (hyperinsulinemia) SUR1MRP8 SURATP-binding cassette transporter sub-family C member 8 TNDM2ATP-binding cassette sub-family C member 8; ATP-binding cassette sub-family C member 8; ATP-binding cassette transporter sub-family C member 8; ATP-binding cassette, sub-family C (CFTR/MRP), member 8; sulfonylurea receptor (hyperinsulinemia); Sulfonylurea receptor 1
Gene Aliases: ABC36; ABCC8; HHF1; HI; HRINS; MRP8; PHHI; SUR; SUR1; SUR1delta2; TNDM2
UniProt ID: (Human) Q09428
Entrez Gene ID: (Human) 6833
Molecular Function: ATP-binding cassette (ABC) transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.