Invitrogen
Manufacturer Code:PA559947
Catalog # PIPA559947
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human SUR1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ABCC8 SUR1 ATP-binding cassette sub-family C (CFTR/MRP) member 8 ABC36 HHF1 HI HRINS MRP8 PHHI SUR TNDM2 ATP-binding cassette sub-family C member 8 sulfonylurea receptor (hyperinsulinemia); ATP-binding cassette sub-family C member 8; ATP-binding cassette transporter sub-family C member 8; ATP-binding cassette, sub-family C (CFTR/MRP), member 8; sulfonylurea receptor (hyperinsulinemia); Sulfonylurea receptor 1
Gene Aliases: ABC36; ABCC8; HHF1; HI; HRINS; MRP8; PHHI; SUR; SUR1; SUR1delta2; TNDM2
UniProt ID: (Human) Q09428
Entrez Gene ID: (Human) 6833
Molecular Function: ATP-binding cassette (ABC) transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.