Invitrogen
Manufacturer Code:PA5103639
Catalog # PIPA5103639
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human ABCC8 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ABC36 Abcc8 AM60008PU-N ATP binding cassette subfamily C member 8 ATP-binding cassette sub-family C member 8 ATP-binding cassette transporter sub-family C member 8 ATP-binding cassette subfamily C (CFTR/MRP) member 8 ATP-binding cassette sub-family C (CFTR/MRP) member 8 D930031B21Rik HHF1 HI HRINS MRP8 PH SUR1 PHHI sulfonylurea receptor sulfonylurea receptor (hyperinsulinemia) sulfonylurea receptor 1 sulphonylurea receptor 1 Sur SUR1 SUR1delta2 TNDM2; ATP-binding cassette sub-family C member 8; ATP-binding cassette transporter sub-family C member 8; ATP-binding cassette, sub-family C (CFTR/MRP), member 8; sulfonylurea receptor; sulfonylurea receptor (hyperinsulinemia); Sulfonylurea receptor 1; sulphonylurea receptor 1
Gene Aliases: ABC36; ABCC8; D930031B21Rik; HHF1; HI; HRINS; MRP8; PHHI; SUR; SUR1; SUR1delta2; TNDM2
UniProt ID: (Human) Q09428, (Rat) Q09429
Entrez Gene ID: (Human) 6833, (Mouse) 20927, (Rat) 25559
Molecular Function: ATP-binding cassette (ABC) transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.