Novus Biologicals
Manufacturer Code:NBP288383
Catalog # NB036005
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the N-terminal region of SUR-8 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KIAA0862FLJ60412 Protein soc-2 homolog Protein sur-8 homolog SIAA0862 SOC-2 soc-2 (suppressor of clear C.elegans) homolog soc-2 suppressor of clear homolog (C. elegans) SOC2SUR8leucine-rich repeat protein SHOC-2 SUR-8; Leucine-rich repeat protein SHOC-2; Protein soc-2 homolog; Protein sur-8 homolog; SHOC2 leucine-rich repeat scaffold protein; soc-2 suppressor of clear homolog
Gene Aliases: KIAA0862; SHOC2; SIAA0862; SOC2; SUR8
UniProt ID: (Human) Q9UQ13
Entrez Gene ID: (Human) 8036
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.