Invitrogen
Manufacturer Code:MA527784
Catalog # PIMA527784
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | GT244 |
Immunogen | Human SUPV3L1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ATP-dependent RNA helicase SUPV3L1, mitochondrial; Suppressor of var1 3-like protein 1; suppressor of var1, 3-like 1; suppressor of var1, 3-like 1(SUV3); SUV3 SUPV3L1 SUV3-like protein 1 suppressor of var1 3-like 1(SUV3) suppressor of var1 3-like protein 1 ATP-dependent RNA helicase SUPV3L1 mitochondrial suppressor of var1 3-like 1 (S. cerevisiae); SUV3-like helicase; SUV3-like protein 1
Gene Aliases: SUPV3L1; SUV3
UniProt ID: (Human) Q8IYB8
Entrez Gene ID: (Human) 6832
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.