Novus Biologicals
Manufacturer Code:NBP183508
Catalog # NBP183508
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:VGYSPMTPGAPSPGGYNPHTPGSGIEQNSSDWVTTDIQVKVRDTYLDTQVVGQTGVIRSVTGGMCSVYLKDSEKVVSISSEHLEPITPTKNNKVKVILGEDREATGVLLSIDGEDGIVRMDLDEQLKILNLRFL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DRB sensitivity-inducing factor 160 kDa subunit; DRB sensitivity-inducing factor 160 kDa subunit DRB sensitivity-inducing factor large subunit DSIF large subunit DSIF p160 FLJ34157 SPT5hSPT5 SPT5HTat-cotransactivator 1 protein suppressor of Ty (S.cerevisiae) 5 homolog suppressor of Ty 5 homolog (S. cerevisiae) Tat-CT1 Tat-CT1 protein transcription elongation factor SPT5; DRB sensitivity-inducing factor large subunit; DSIF large subunit; DSIF p160; hSPT5; suppressor of Ty 5 homolog; Tat-cotransactivator 1 protein; Tat-CT1 protein; Transcription elongation factor SPT5
Gene Aliases: SPT5; SPT5H; SUPT5H; Tat-CT1
UniProt ID: (Human) O00267
Entrez Gene ID: (Human) 6829
Molecular Function:
DNA binding protein
chromatin/chromatin-binding protein
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.