Novus Biologicals
Manufacturer Code:NB11040592
Catalog # NB11040592
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residue 50 and 100 of human Suppressor of Ty 5 Homolog using the numbering given in entry NP_003160.2 (GeneID 6829). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DRB sensitivity-inducing factor 160 kDa subunit; DRB sensitivity-inducing factor 160 kDa subunit DRB sensitivity-inducing factor large subunit DSIF large subunit DSIF p160 FLJ34157 SPT5hSPT5 SPT5HTat-cotransactivator 1 protein suppressor of Ty (S.cerevisiae) 5 homolog suppressor of Ty 5 homolog (S. cerevisiae) Tat-CT1 Tat-CT1 protein transcription elongation factor SPT5; DRB sensitivity-inducing factor large subunit; DSIF large subunit; DSIF p160; hSPT5; suppressor of Ty 5 homolog; Tat-cotransactivator 1 protein; Tat-CT1 protein; Transcription elongation factor SPT5
Gene Aliases: SPT5; SPT5H; SUPT5H; Tat-CT1
UniProt ID: (Human) O00267
Entrez Gene ID: (Human) 6829
Molecular Function:
DNA binding protein
chromatin/chromatin-binding protein
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.