Invitrogen
Manufacturer Code:PA5103308
Catalog # PIPA5103308
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human SPT4H corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AL022777 DRB sensitivity-inducing factor 14 kDa subunit DRB sensitivity-inducing factor small subunit DRB sensitivity-inducing factor small subunit 1 DSIF p14 DSIF small subunit DSIF small subunit 1 hSPT4 small hSpt4 subunit SPT4 SPT4 homolog DSIF elongation factor subunit SPT4H suppressor of Ty 4 homolog 1 suppressor of Ty 4 homolog 1 (S. cerevisiae) suppressor of Ty 4 homolog 2 suppressor of Ty 4A Supt4a SUPT4H SUPT4H1 Supt4h1a Supt4h2 Transcription elongation factor SPT4 transcription elongation factor SPT4 1 Transcription elongation factor SPT4-A; DRB sensitivity-inducing factor 14 kDa subunit; DRB sensitivity-inducing factor small subunit; DRB sensitivity-inducing factor small subunit 1; DSIF p14; DSIF small subunit; DSIF small subunit 1; hSPT4; small hSpt4 subunit; suppressor of Ty 4 homolog 1; Transcription elongation factor SPT4; Transcription elongation factor SPT4 1; Transcription elongation factor SPT4-A
Gene Aliases: AL022777; SPT4; SPT4H; Supt4a; SUPT4H; SUPT4H1; Supt4h1a
UniProt ID: (Human) P63272, (Mouse) P63271
Entrez Gene ID: (Human) 6827, (Mouse) 20922
Molecular Function: DNA binding protein chromatin/chromatin-binding protein nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.