Abnova
Manufacturer Code:MAB12948
Catalog # 89-934-978
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 8D2 |
Immunogen | Recombinant protein corresponding to full length human SUPT16H. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. Do not freeze. |
Mouse monoclonal antibody raised against full length recombinant human SUPT16H.
For Research Use Only
Protein Aliases: CDC68; Chromatin-specific transcription elongation factor 140 kDa subunit; facilitates chromatin remodeling 140 kDa subunit; Facilitates chromatin transcription complex subunit SPT16; FACT; FACT 140 kDa subunit; FACT complex subunit SPT16; FACTP140; FLJ10857; FLJ14010; FLJ34357; hSPT16; SPT16/CDC68; suppressor of Ty 16 homolog
Gene Aliases: CDC68; FACT140; FACTP140; SPT16; SPT16/CDC68; SUPT16H
UniProt ID: (Human) Q9Y5B9
Entrez Gene ID: (Human) 11198
Molecular Function: nucleic acid binding transcription cofactor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.