Invitrogen
Manufacturer Code:PA540388
Catalog # PIPA540388
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human SUPT16H |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Chromatin-specific transcription elongation factor 140 kDa subunit; chromatin-specific transcription elongation factor large subunit; DNA unwinding factor 140 kDa subunit; facilitates chromatin remodeling 140 kDa subunit; Facilitates chromatin transcription complex subunit SPT16; FACT 140 kDa subunit; FACT complex subunit SPT16; FACT complex subunit SPT16-like protein; FACTp140; hSPT16; suppressor of Ty 16 homolog; Suppressor of Ty 16 homolog (S. cerevisiae) CDC68 FACT FACTP140 FLJ10857 FLJ14010 FLJ34357 SPT16/CDC68
Gene Aliases: CDC68; DUF140; FACT140; FACTP140; fq44d08; SPT16; SPT16/CDC68; Supt16; SUPT16H; wu:fq44d08; zgc:194708; zgc:194732
UniProt ID: (Human) Q9Y5B9, (Mouse) Q920B9
Entrez Gene ID: (Human) 11198, (Pig) 100155525, (Bovine) 514989, (Rabbit) 100353045, (Horse) 100059304, (Dog) 612874, (Mouse) 114741, (Rat) 305851, (Guinea pig) 100718509, (Zebrafish) 562348
Molecular Function: nucleic acid binding transcription cofactor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.