Invitrogen
Manufacturer Code:PA518443
Catalog # PIPA518443
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (DLTNKEGKKPEEK) corresponding to the internal amino acids of SUPT16H |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Chromatin-specific transcription elongation factor 140 kDa subunit; facilitates chromatin remodeling 140 kDa subunit; Facilitates chromatin transcription complex subunit SPT16; FACT 140 kDa subunit; FACT complex subunit SPT16; FACTp140; hSPT16; suppressor of Ty 16 homolog; SUPT16H FACT suppressor of Ty 16 homolog (S. cerevisiae) HGNC:11465 CDC68 FACTP140 FLJ10857 FLJ14010 SPT16/CDC68 chromatin-specific transcription elongation factor large subunit FLJ34357 facilitates chromatin remodeling 140 kDa subunit
Gene Aliases: CDC68; FACT140; FACTP140; SPT16; SPT16/CDC68; SUPT16H
UniProt ID: (Human) Q9Y5B9
Entrez Gene ID: (Human) 11198
Molecular Function: nucleic acid binding transcription cofactor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.