Abnova
Manufacturer Code:PAB15578
Catalog # 89-117-652
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Baker's yeast |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids 494-507 of <i>Saccharomyces cerevisiae</i> SUP35. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of SUP35.
For Research Use Only
Protein Aliases: ERF-3; ERF2; ERF3; Eukaryotic peptide chain release factor GTP-binding subunit; G1 to S phase transition protein 1; GST1; Omnipotent suppressor protein 2; PNM2; Polypeptide release factor 3; PSI no more protein 2; SAL3; SUF12; SUP2; SUP36; Translation release factor 3; translation termination factor GTPase eRF3
Gene Aliases: GST1; PNM2; SAL3; SUF12; SUP2; SUP35; SUP36; YD9395.05; YDR172W
UniProt ID: (Baker's yeast) P05453
Entrez Gene ID: (Baker's yeast) 851752
Molecular Function:
G-protein
RNA binding protein
enzyme modulator
hydrolase
nucleic acid binding
translation elongation factor
translation factor
translation initiation factor
translation release factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.