Novus Biologicals
Manufacturer Code:NBP274411AF405
Catalog # N274411A405
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI5A2 |
Immunogen | Recombinant protein fragment corresponding to amino acids 320-545 of human SUOX (NP_000447) produced in E.coli. |
Conjugate | Alexa Fluor 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: EC 1.8.3.1 sulfite oxidase sulfite oxidase mitochondrial; Sulfite oxidase, mitochondrial
Gene Aliases: SUOX
UniProt ID: (Human) P51687
Entrez Gene ID: (Human) 6821
Molecular Function:
oxidase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.