Novus Biologicals
Manufacturer Code:NBP15541320UL
Catalog # NBP15541320
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to SUNC1(Sad1 and UNC84 domain containing 1) The peptide sequence was selected from the C terminal of SUNC1. Peptide sequence IKLATKIIPTAVTMEHISEKVSPSGNISSAPKEFSVYGITKKCEGEEIFL. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hypothetical protein MGC33329; MGC33329 Sad1 and UNC84 domain containing 1 Sad1 and UNC84 domain containing 3 Sad1/unc-84 domain-containing protein 1 SUN domain-containing protein 3 SUNC1; Sad1 and UNC84 domain containing 1; Sad1/unc-84 domain-containing protein 1; SUN domain-containing protein 3
Gene Aliases: SUN3; SUNC1
UniProt ID: (Human) Q8TAQ9
Entrez Gene ID: (Human) 256979
Molecular Function: cytoskeletal protein microtubule family cytoskeletal protein non-motor microtubule binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.