Proteintech
Manufacturer Code:264341AP150UL
Catalog # 50-173-5338
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SUNC1 Fusion Protein Ag23780 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hypothetical protein MGC33329; Sad1 and UNC84 domain containing 1; Sad1/unc-84 domain-containing protein 1; SUN domain-containing protein 3; SUN3 SUNC1
Gene Aliases: SUN3; SUNC1
UniProt ID: (Human) Q8TAQ9, (Mouse) Q5SS91
Entrez Gene ID: (Human) 256979, (Mouse) 194974
Molecular Function:
cytoskeletal protein
microtubule family cytoskeletal protein
non-motor microtubule binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.