Novus Biologicals
Manufacturer Code:NBP187395
Catalog # NBP187395
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:QGNFFSFLPVLNWASMHRTQRVDDPQDVFKPTTSRLKQPLQGDSEAFPWHWMSGVEQQVASLSGQCHHHGENLRELTTLLQKLQARVDQMEGGAAGPSASVRDAVGQPPRETDFMAFHQEHEVRMSHLEDILGKLR |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KIAA0810UNC84AFLJ12407 MGC176649 Protein unc-84 homolog A Sad1 and UNC84 domain containing 1 Sad1 unc-84 domain protein 1 Sad1/unc-84 protein-like 1 SUN domain-containing protein 1 unc-84 homolog A unc-84 homolog A (C. elegans); Protein unc-84 homolog A; Sad1 unc-84 domain protein 1; Sad1/unc-84 protein-like 1; SUN domain-containing protein 1; unc-84 homolog A
Gene Aliases: KIAA0810; SUN1; UNC84A
UniProt ID: (Human) O94901
Entrez Gene ID: (Human) 23353
Molecular Function:
cytoskeletal protein
microtubule family cytoskeletal protein
non-motor microtubule binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.