Novus Biologicals
Manufacturer Code:NBP267419
Catalog # NBP267419
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ChIP assay (ChIP) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | SJ20-03 |
Immunogen | Synthetic peptide within C-terminal human SUMO-1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
Protein Aliases: DAP1 GAP modifying protein 1 GAP-modifying protein 1 GMP1SMT3CSMT3H3OFC10UBL1PIC1 SENP2 sentrin small ubiquitin-related modifier 1 SMT3 SMT3 homolog 3 SMT3 suppressor of mif two 3 homolog 1 (S. cerevisiae) SMT3 suppressor of mif two 3 homolog 1 (yeast) Smt3C SUMO-1 Ubiquitin-homology domain protein PIC1 ubiquitin-like 1 (sentrin) Ubiquitin-like protein SMT3C Ubiquitin-like protein UBL1; GAP modifying protein 1; GAP-modifying protein 1; GMP1; Sentrin; Small ubiquitin-related modifier 1; SMT3 homolog 3; SMT3 suppressor of mif two 3 homolog 1; Smt3C; SUMO-1; Ubiquitin-homology domain protein PIC1; Ubiquitin-like protein SMT3C; Ubiquitin-like protein UBL1
Gene Aliases: DAP1; GMP1; OFC10; OK/SW-cl.43; PIC1; SENP2; SMT3; SMT3C; SMT3H3; SUMO1; UBL1
UniProt ID: (Human) P63165
Entrez Gene ID: (Human) 7341
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.