Invitrogen
Manufacturer Code:PA511312
Catalog # PIPA511312
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 55-86 amino acids from the C-terminal region of human SUMO1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GAP modifying protein 1; GAP-modifying protein 1; GMP1; Sentrin; Small ubiquitin-related modifier 1; Small ubiquitin-related modifier 1 SUMO-1 Ubiquitin-like protein SMT3C SMT3 homolog 3 Ubiquitin-homology domain protein PIC1 Ubiquitin-like protein UBL1 GAP modifying protein 1 GMP1 Sentrin SMT3C SMT3H3 UBL1; SMT3 homolog 3; SMT3 suppressor of mif two 3 homolog 1; Smt3C; SUMO-1; Ubiquitin-homology domain protein PIC1; Ubiquitin-like protein SMT3C; Ubiquitin-like protein UBL1
Gene Aliases: DAP1; GMP1; OFC10; OK/SW-cl.43; PIC1; SENP2; SMT3; SMT3C; SMT3H3; SUMO1; UBL1
UniProt ID: (Human) P63165
Entrez Gene ID: (Human) 7341
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.