Proteintech
Manufacturer Code:103291AP150UL
Catalog # 50-172-6218
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SUMO1 Fusion Protein Ag0414 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DAP 1 GAP modifying protein 1 GMP1 OFC10 PIC1 SENP2 Sentrin SMT3 SMT3 homolog 3 SMT3C SMT3H3 SUMO 1 SUMO1 Ubiquitin like protein SMT3C Ubiquitin like protein UBL1 UBL1; GAP modifying protein 1; GAP-modifying protein 1; GMP1; Sentrin; Small ubiquitin-related modifier 1; small ubiquitin-related modifier-1; SMT3 homolog 3; SMT3 suppressor of mif two 3 homolog 1; Smt3C; SUMO-1; SUMO-1 related peptidase; Ubiquitin-homology domain protein PIC1; ubiquitin-like 1; ubiquitin-like 1 (sentrin); Ubiquitin-like protein SMT3C; Ubiquitin-like protein UBL1
Gene Aliases: DAP1; GMP1; OFC10; OK/SW-cl.43; PIC1; SENP2; SENTRIN; SMT3; SMT3C; SMT3H3; SMTP3; SUMO-1; SUMO1; UBL1
UniProt ID: (Human) P63165, (Mouse) P63166, (Rat) Q5I0H3
Entrez Gene ID: (Human) 7341, (Mouse) 22218, (Rat) 301442
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.