Novus Biologicals
Manufacturer Code:NBP230700
Catalog # NBP230700
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to amino acids: MKIQQLHPANAKTVEWDWKLLTYVMEEEGQTLPGRVLFLRYVVQTLEDDFQQTLRRQRQHLQQSIANMVLSCDKQPHNVRDVIKWLVKA |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Chromosome 5 Open Reading Frame 25 Oocyte Maturation Associated 1 OOMA1 Platform Element For Inhibition Of Autolytic Degradation PLEIAD SIMC1 SUMO-Interacting Motif-Containing Protein 1 SUMO-Interacting Motifs Containing 1; oocyte maturation associated 1; Platform element for inhibition of autolytic degradation; SUMO-interacting motif-containing protein 1; SUMO-interacting motifs containing 1
Gene Aliases: C5orf25; OOMA1; PLEIAD; SIMC1
UniProt ID: (Human) Q8NDZ2
Entrez Gene ID: (Human) 375484
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.