Novus Biologicals
Manufacturer Code:NBP274410AF405
Catalog # N274410A405
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI6B11 |
Immunogen | Full length recombinant protein of human SUMO Activating Enzyme E1 (SAE1) (NP_005491) produced in HEK293T cells. |
Conjugate | Alexa Fluor 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: activator of SUMO1; AOS1activator of SUMO1 FLJ3091 HSPC140 Sua1 SUA1sentrin/SUMO-activating protein AOS1 SUMO-1 activating enzyme E1 N subunit SUMO1 activating enzyme subunit 1 SUMO-1 activating enzyme subunit 1 SUMO-activating enzyme subunit 1 Ubiquitin-like 1-activating enzyme E1A ubiquitin-like protein SUMO-1 activating enzyme UBLE1A; sentrin/SUMO-activating protein AOS1; SUMO-1 activating enzyme E1 N subunit; SUMO-1 activating enzyme subunit 1; SUMO-activating enzyme subunit 1; SUMO-activating enzyme subunit 1, N-terminally processed; Ubiquitin-like 1-activating enzyme E1A; ubiquitin-like protein SUMO-1 activating enzyme
Gene Aliases: AOS1; HSPC140; SAE1; SUA1; UBLE1A
UniProt ID: (Human) Q9UBE0
Entrez Gene ID: (Human) 10055
Molecular Function: ligase transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.