Proteintech
Manufacturer Code:112101AP150UL
Catalog # 50-172-6971
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SUMF2 Fusion Protein Ag1711 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C-alpha-formyglycine-generating enzyme 2; C-alpha-formylglycine-generating enzyme 2; Inactive C-alpha-formylglycine-generating enzyme 2; Paralog of formylglycine-generating enzyme; paralog of the formylglycine-generating enzyme; pFGE; pFGE sulfatase modifying factor 2 SUMF2 UNQ1968/PRO4500; Sulfatase-modifying factor 2
Gene Aliases: 2610040F05Rik; pFGE; PSEC0171; SUMF2; UNQ1968/PRO4500
UniProt ID: (Human) B4DWQ0, (Mouse) Q8BPG6
Entrez Gene ID: (Human) 25870, (Mouse) 67902, (Rat) 360800
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.