Proteintech
Manufacturer Code:125781AP150UL
Catalog # 50-172-8081
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SULT4A1 Fusion Protein Ag3271 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BR STL 1 BRSTL1 hBR STL hBR STL 1 NST ST4A1 Sulfotransferase 4A1 SULT4A1 SULTX3; Brain sulfotransferase-like protein; hBR-STL; mBR-STL; nervous system cytosolic sulfotransferase; Nervous system sulfotransferase; NST; rBR-STL; ST4A1; Sulfotransferase 4A1; sulfotransferase family 4A, member 1; sulfotransferase-related protein
Gene Aliases: 2400007A17Rik; AI853543; BR-STL-1; BRSTL1; DJ388M5.3; hBR-STL-1; NST; ST4A1; SULT4A1; SULTX3
UniProt ID: (Human) Q9BR01, (Mouse) P63046, (Rat) P63047
Entrez Gene ID: (Human) 25830, (Mouse) 29859, (Rat) 58953
Molecular Function: transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.