Invitrogen
Manufacturer Code:MA537788
Catalog # PIMA537788
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2142CT1553.26.63 |
Immunogen | reconbinant protein from human SULT2B1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: AI326997 Alcohol sulfotransferase BB173635 HSST2 Hydroxysteroid sulfotransferase 2 RGD:1308882} ST2B1 sulfotransferase 2B sulfotransferase 2B1 Sulfotransferase family 2B member 1 sulfotransferase family cytosolic 2B member 1 sulfotransferase family cytosolic 2B member 1 Sult2b SULT2B1 sult2b1 {ECO:0000312 SULT2B1a; Alcohol sulfotransferase; Hydroxysteroid sulfotransferase 2; ST2B1; Sulfotransferase 2B1; Sulfotransferase family 2B member 1; Sulfotransferase family cytosolic 2B member 1; sulfotransferase family, cytosolic, 2B, member 1
Gene Aliases: HSST2; SULT2B1
UniProt ID: (Human) O00204
Entrez Gene ID: (Human) 6820
Molecular Function: transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.