Invitrogen
Manufacturer Code:PA587025
Catalog # PIPA587025
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein of human SULT2B1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alcohol sulfotransferase; HSST2; Hydroxysteroid sulfotransferase 2; ST2B1; sulfotransferase 2B; Sulfotransferase 2B1; Sulfotransferase family 2B member 1; Sulfotransferase family cytosolic 2B member 1; sulfotransferase family, cytosolic, 2B, member 1; sult2b1 {ECO:0000312|RGD:1308882}
Gene Aliases: AI326997; BB173635; HSST2; ST2B1; SULT2B; SULT2B1
UniProt ID: (Human) O00204, (Mouse) O35400, (Rat) Q29YR5
Entrez Gene ID: (Human) 6820, (Mouse) 54200, (Rat) 292915
Molecular Function: transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.