Invitrogen
Manufacturer Code:PA580085
Catalog # PIPA580085
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence in the middle region of human SULT2B1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alcohol sulfotransferase; Hydroxysteroid sulfotransferase 2; ST2B1; sulfotransferase 2B; Sulfotransferase 2B1; Sulfotransferase family 2B member 1; Sulfotransferase family cytosolic 2B member 1; Sulfotransferase family cytosolic 2B member 1 ST2B1 Sulfotransferase 2B1 Alcohol sulfotransferase Hydroxysteroid sulfotransferase 2 SULT2B1 HSST2; sulfotransferase family, cytosolic, 2B, member 1; sult2b1 {ECO:0000312|RGD:1308882}
Gene Aliases: HSST2; ST2B1; SULT2B1
UniProt ID: (Human) O00204, (Rat) Q29YR5
Entrez Gene ID: (Human) 6820, (Rat) 292915
Molecular Function: transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.