OriGene
Manufacturer Code:CF501658
Catalog # 50-166-0654
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2A6 |
Immunogen | Full length human recombit protein of human SULT2A1 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: alcohol/hydroxysteroid sulfotransferase; Bile salt sulfotransferase; bile-salt sulfotranasferase 2A1; Dehydroepiandrosterone sulfotransferase; DHEA-ST; DHEA-ST DHEAS HST ST2 ST2A1 ST2A3 STD hSTa; HST; Hydroxysteroid Sulfotransferase; ST2; ST2A1; Sulfotransferase 2A1; sulfotransferase family, cytosolic, 2A, dehydroepiandrosterone (DHEA)-preferring, member 1; SULT2A3
Gene Aliases: DHEA-ST; DHEAS; HST; hSTa; ST2; ST2A1; ST2A3; STD; SULT2A1
UniProt ID: (Human) Q06520
Entrez Gene ID: (Human) 6822
Molecular Function: transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.