Invitrogen
Manufacturer Code:PA512243
Catalog # PIPA512243
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 253-285 amino acids from the C-terminal region of human SULT2A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alcohol sulfotransferase Hydroxysteroid Sulfotransferase HST Dehydroepiandrosterone sulfotransferase DHEA-ST ST2 ST2A3 HST STD; alcohol/hydroxysteroid sulfotransferase; Bile salt sulfotransferase; Bile salt sulfotransferase 1; bile-salt sulfotranasferase 2A1; Dehydroepiandrosterone sulfotransferase; DHEA-ST; HST; Hydroxysteroid Sulfotransferase; ST; ST2; ST2A1; Sulfotransferase 2A1; sulfotransferase family, cytosolic, 2A, dehydroepiandrosterone (DHEA)-preferring, member 1; sulfotransferase, DHEA preferring; sulfotransferase, hydroxysteroid preferring 1; SULT2A3
Gene Aliases: DHEA-ST; DHEAS; HST; hSTa; mSTa1; ST2; ST2A1; ST2A3; Sta1; STD; Sth1; SULT2A1
UniProt ID: (Human) Q06520, (Mouse) P52843
Entrez Gene ID: (Human) 6822, (Mouse) 20859
Molecular Function: transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.