Invitrogen
Manufacturer Code:PA577218
Catalog # PIPA577218
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human SULT1E1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EST-2; EST-3; EST-6; estrogen sulfotransferase; Estrogen sulfotransferase EST-1 Sulfotransferase 1E1 ST1E1 Sulfotransferase estrogen-preferring SULT1E1 STE; Estrogen sulfotransferase, isoform 3; estrogen sulfotransferase, testis isoform; Estrone sulfotransferase; sulfotransferase 1E1; sulfotransferase, estrogen preferring; Sulfotransferase, estrogen-preferring
Gene Aliases: EST; ST1E1; Ste
UniProt ID: (Rat) P49889
Entrez Gene ID: (Mouse) 20860, (Rat) 360268
Molecular Function: transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.