Proteintech
Manufacturer Code:125221AP150UL
Catalog # 50-172-8039
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SULT1E1 Fusion Protein Ag3216 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EST EST 1 Estrogen sulfotransferase ST1E1 STE Sulfotransferase 1E1 SULT1E1; EST-1; EST-2; EST-3; EST-6; Estrogen sulfotransferase; Estrogen sulfotransferase, isoform 3; estrogen sulfotransferase, testis isoform; Estrone sulfotransferase; ST1E1; Sulfotransferase 1E1; sulfotransferase family 1E, estrogen-preferring, member 1; sulfotransferase, estrogen preferring; Sulfotransferase, estrogen-preferring
Gene Aliases: EST; EST-1; ST1E1; STE; SULT1E1
UniProt ID: (Human) P49888, (Rat) P49889
Entrez Gene ID: (Human) 6783, (Mouse) 20860, (Rat) 360268
Molecular Function: transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.