Invitrogen
Manufacturer Code:PA524548
Catalog # PIPA524548
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 264-292 amino acids from the C-terminal region of human SULT1C3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ST1C3; ST1C3 sulfotransferase 1C3; Sulfotransferase 1C3; sulfotransferase family, cytosolic, 1C, member 3
Gene Aliases: ST1C3; SULT1C3
UniProt ID: (Human) Q6IMI6
Entrez Gene ID: (Human) 442038
Molecular Function: transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.