Invitrogen
Manufacturer Code:PA5112731
Catalog # PIPA5112731
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Sulfotransferase family cytosolic 1B member 1 protein (1-271AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DOPA/tyrosine sulfotransferase; ST1B1; ST1B1 ST1B2 sulfotransferase 1B1 Sulfotransferase 1B2 sulfotransferase family 1B member 1 Sulfotransferase family cytosolic 1B member 1 sulfotransferase family cytosolic 1B member 1 SULT1B1 SULT1B2 Thyroid hormone sulfotransferase; ST1B2; sulfotransferase 1B1; Sulfotransferase 1B2; Sulfotransferase family cytosolic 1B member 1; sulfotransferase family, cytosolic, 1B, member 1; Thyroid hormone sulfotransferase
Gene Aliases: ST1B1; ST1B2; St2b2; SULT1B1; SULT1B2
UniProt ID: (Human) O43704, (Mouse) Q9QWG7
Entrez Gene ID: (Human) 27284, (Mouse) 56362
Molecular Function:
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.