Invitrogen
Manufacturer Code:PA562304
Catalog # PIPA562304
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human SULT1A2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Aryl sulfotransferase 2; arylamine sulfotransferase; P-PST 2; Phenol sulfotransferase 2; phenol-preferring phenol sulfotransferase2; Phenol-sulfating phenol sulfotransferase 2; phenolic-metabolizing (P) form of PST; ST1A2; Sulfotransferase 1A2; Sulfotransferase 1A2 ST1A2 Aryl sulfotransferase 2 Phenol sulfotransferase 2 Phenol-sulfating phenol sulfotransferase 2 P-PST 2 SULT1A2 STP2; sulfotransferase family, cytosolic, 1A, phenol-preferring, member 2; thermostable phenol sulfotransferase
Gene Aliases: HAST4; P-PST; ST1A2; STP2; SULT1A2; TSPST2
UniProt ID: (Human) P50226
Entrez Gene ID: (Human) 6799
Molecular Function:
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.