Invitrogen
Manufacturer Code:MA529589
Catalog # PIMA529589
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3C5H6 |
Immunogen | Recombit Human SULT1A1 protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Aryl sulfotransferase 1; aryl sulfotransferase Catecholamine Sulfating Phenol Sulfotransferase CSPS; HAST1/HAST2; P-PST 1; Phenol sulfotransferase 1; Phenol-sulfating phenol sulfotransferase 1; ST1A1; ST1A3; Sulfotransferase 1A1; sulfotransferase family, cytosolic, 1A, phenol-preferring, member 1; Thermostable phenol sulfotransferase; thermostable phenol sulfotransferase1; Ts-PST
Gene Aliases: HAST1/HAST2; OK/SW-cl.88; P-PST; PST; ST1A1; ST1A3; STP; STP1; SULT1A1; TSPST1
UniProt ID: (Human) P50225
Entrez Gene ID: (Human) 6817
Molecular Function: transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.