Proteintech
Manufacturer Code:122601AP150UL
Catalog # 50-172-7840
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SULF2 Fusion Protein Ag2915 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Extracellular sulfatase Sulf 2 HSULF 2 KIAA1247 SULF2 sulfatase 2; Extracellular sulfatase Sulf-2; hSulf-2; mSulf-2; sulfatase FP2
Gene Aliases: 2010004N24Rik; AU020235; HSULF-2; KIAA1247; mKIAA1247; MSulf-2; SULF2; UNQ559/PRO1120
UniProt ID: (Human) Q8IWU5, (Mouse) Q8CFG0
Entrez Gene ID: (Human) 55959, (Rat) 311642, (Mouse) 72043
Molecular Function:
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.